C12H316CE988 Chemcia Scientific, LLC

6-Fluoro-3-iodo-imidazo[1,2-a]pyridine

  • MDL number MFCD20483025
  • Cas Number : 1219127-01-1
  • Molecular Weight : 262.02613

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2443

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2443
Availability details for
Quantity
  • Availability
  • Add to Cart