C12H316CE989 Chemcia Scientific, LLC

8-Bromo-3-iodo-imidazo[1,2-a]pyridine

  • MDL number MFCD22627990
  • Cas Number : 1232038-69-5
  • Molecular Weight : 322.93173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Greenish Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2444

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2444
Availability details for
Quantity
  • Availability
  • Add to Cart