C12H316CE98D Chemcia Scientific, LLC

2-Bromo-8-methoxy-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD22571379
  • Cas Number : 1319067-40-7
  • Molecular Weight : 228.04937

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2477

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2477
Availability details for
Quantity
  • Availability
  • Add to Cart