C12H316CE993 Chemcia Scientific, LLC

2-Chloro-6,7-dihydro-4H-thiazolo[5,4-c]pyridine-5-carboxylic acid tert-butyl ester

  • MDL number MFCD22381608
  • Cas Number : 1221931-40-3
  • Molecular Weight : 274.7714

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2504

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2504
Availability details for
Quantity
  • Availability
  • Add to Cart