C12H316CE995 Chemcia Scientific, LLC

2-Bromo-benzothiazol-6-ylamine

  • MDL number MFCD11977496
  • Cas Number : 945400-80-6
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1,3-benzothiazol-6-amine
Inchi Code 1S/C7H5BrN2S/c8-7-10-5-2-1-4(9)3-6(5)11-7/h1-3H,9H2
Inchi Key PKRUNKROSSHAGG-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2508

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2508
Availability details for
Quantity
  • Availability
  • Add to Cart