C12H316CE997 Chemcia Scientific, LLC

1-Chloro-4-cyclopropoxy-benzene

  • MDL number MFCD17000492
  • Cas Number : 17204-68-1
  • Molecular Weight : 168.62448

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2519

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2519
Availability details for
Quantity
  • Availability
  • Add to Cart