C12H316CE99F Chemcia Scientific, LLC

6-Benzyloxy-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid ethyl ester

  • MDL number MFCD27918562
  • Cas Number : 869358-02-1
  • Molecular Weight : 323.35164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2551

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2551
Availability details for
Quantity
  • Availability
  • Add to Cart