C12H316CE9A1 Chemcia Scientific, LLC

6-Benzyloxy-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid

  • MDL number MFCD19298538
  • Cas Number : 57278-54-3
  • Molecular Weight : 295.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(benzyloxy)-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid
Inchi Code 1S/C17H13NO4/c19-16-13-8-12(22-10-11-4-2-1-3-5-11)6-7-15(13)18-9-14(16)17(20)21/h1-9H,10H2,(H,18,19)(H,20,21)
Inchi Key BJZQPNHESNGXAR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2553

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2553
Availability details for
Quantity
  • Availability
  • Add to Cart