C12H316CE9A3 Chemcia Scientific, LLC

6-Cyano-4-oxo-1,4-dihydro-quinoline-3-carboxylic acid ethyl ester

  • MDL number MFCD10000762
  • Molecular Weight : 242.23625

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2569

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2569
Availability details for
Quantity
  • Availability
  • Add to Cart