C12H316CE9A8 Chemcia Scientific, LLC

1-Bromo-4-cyclopentyloxy-benzene

  • MDL number MFCD08461672
  • Cas Number : 30752-30-8
  • Molecular Weight : 241.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-4-(cyclopentyloxy)benzene
Inchi Code 1S/C11H13BrO/c12-9-5-7-11(8-6-9)13-10-3-1-2-4-10/h5-8,10H,1-4H2
Inchi Key DWMSXAAIGFYBGC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2580

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2580
Availability details for
Quantity
  • Availability
  • Add to Cart