C12H316CE9AB Chemcia Scientific, LLC

1-Chloro-3-cyclopropoxy-benzene

  • MDL number MFCD17000494
  • Cas Number : 1243285-60-0
  • Molecular Weight : 168.62448

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Colorless Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2585

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2585
Availability details for
Quantity
  • Availability
  • Add to Cart