C12H316CE9AE Chemcia Scientific, LLC

3-Isopropoxy-benzoic acid methyl ester

  • MDL number MFCD01922057
  • Cas Number : 350989-42-3
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-isopropoxybenzoate
Inchi Code 1S/C11H14O3/c1-8(2)14-10-6-4-5-9(7-10)11(12)13-3/h4-8H,1-3H3
Inchi Key GKWWRXCPQAQURQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2593

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2593
Availability details for
Quantity
  • Availability
  • Add to Cart