C12H316CE9B0 Chemcia Scientific, LLC

3-Cyclopropoxy-benzoic acid

  • MDL number MFCD17000471
  • Cas Number : 1211587-15-3
  • Molecular Weight : 178.1894

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2597

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2597
Availability details for
Quantity
  • Availability
  • Add to Cart