C12H316CE9B2 Chemcia Scientific, LLC

3-Cyclobutoxy-benzoic acid

  • MDL number MFCD22989041
  • Cas Number : 1260113-15-2
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyclobutoxybenzoic acid
Inchi Code 1S/C11H12O3/c12-11(13)8-3-1-6-10(7-8)14-9-4-2-5-9/h1,3,6-7,9H,2,4-5H2,(H,12,13)
Inchi Key LHXRYGWBPVPGFU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2600

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2600
Availability details for
Quantity
  • Availability
  • Add to Cart