C12H316CE9B7 Chemcia Scientific, LLC

1-Bromo-2-cyclopentyloxy-benzene

  • MDL number MFCD09950866
  • Cas Number : 494773-69-2
  • Molecular Weight : 241.12966

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Colorless Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2631

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2631
Availability details for
Quantity
  • Availability
  • Add to Cart