C12H316CE9BD Chemcia Scientific, LLC

1-Cyclopropoxy-4-fluoro-benzene

  • MDL number MFCD17000409
  • Cas Number : 1205501-33-2
  • Molecular Weight : 152.16988

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Colorless Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2652

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2652
Availability details for
Quantity
  • Availability
  • Add to Cart