C12H316CE9C7 Chemcia Scientific, LLC

5-Chloro-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09836168
  • Cas Number : 1000017-93-5
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Inchi Code 1S/C8H5ClN2O2/c9-6-2-1-3-7-10-5(8(12)13)4-11(6)7/h1-4H,(H,12,13)
Inchi Key TVUHAHMCORHJBN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2678

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2678
Availability details for
Quantity
  • Availability
  • Add to Cart