C12H316CE9C9 Chemcia Scientific, LLC

5-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD11870491
  • Cas Number : 175965-64-7
  • Molecular Weight : 168.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro[1,2,4]triazolo[1,5-a]pyridin-2-ylamine
Inchi Code 1S/C6H5ClN4/c7-4-2-1-3-5-9-6(8)10-11(4)5/h1-3H,(H2,8,10)
Inchi Key RCCCLZDQZNFDGK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2681

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2681
Availability details for
Quantity
  • Availability
  • Add to Cart