C12H316CEA21 Chemcia Scientific, LLC

5-Chloro-3-methoxy-pyridin-2-ylamine

  • MDL number MFCD17015845
  • Cas Number : 1242336-53-3
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-methoxy-2-pyridinamine
Inchi Code 1S/C6H7ClN2O/c1-10-5-2-4(7)3-9-6(5)8/h2-3H,1H3,(H2,8,9)
Inchi Key CKFFRODMHNJVSQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2713

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2713
Availability details for
Quantity
  • Availability
  • Add to Cart