C12H316CEA24 Chemcia Scientific, LLC

(4-Chloro-2-trifluoromethyl-phenyl)-thiourea

  • MDL number MFCD06763403
  • Cas Number : 648415-77-4
  • Molecular Weight : 254.66262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2743

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2743
Availability details for
Quantity
  • Availability
  • Add to Cart