C12H316CEA25 Chemcia Scientific, LLC

4-Iodo-5-methoxy-2-nitro-phenylamine

  • MDL number MFCD22570398
  • Cas Number : 335349-66-1
  • Molecular Weight : 294.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-iodo-5-methoxy-2-nitroaniline
Inchi Code 1S/C7H7IN2O3/c1-13-7-3-5(9)6(10(11)12)2-4(7)8/h2-3H,9H2,1H3
Inchi Key LFEDHUMOJYMWQN-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2764

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2764
Availability details for
Quantity
  • Availability
  • Add to Cart