C12H316CEA2D Chemcia Scientific, LLC

2-(4-Bromo-phenyl)-3-methyl-butyric acid

  • MDL number MFCD11557244
  • Cas Number : 51632-30-5
  • Molecular Weight : 257.12906

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2693

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2693
Availability details for
Quantity
  • Availability
  • Add to Cart