C12H316CEA2E Chemcia Scientific, LLC

2-(4-Bromo-phenyl)-3-methyl-butyric acid methyl ester

  • MDL number MFCD16340133
  • Cas Number : 1061284-70-5
  • Molecular Weight : 271.15615

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Colorless Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2697

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2697
Availability details for
Quantity
  • Availability
  • Add to Cart