C12H316CEA30 Chemcia Scientific, LLC

1-(4-Bromo-phenyl)-cyclopropanecarboxylic acid methyl ester

  • MDL number MFCD12913964
  • Cas Number : 638220-35-6
  • Molecular Weight : 255.11312

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Low MP Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2700

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2700
Availability details for
Quantity
  • Availability
  • Add to Cart