C12H316CEA31 Chemcia Scientific, LLC

1-(4-Bromo-phenyl)-cyclopentanecarboxylic acid methyl ester

  • MDL number MFCD18711571
  • Cas Number : 1236357-64-4
  • Molecular Weight : 283.1673

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2702

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2702
Availability details for
Quantity
  • Availability
  • Add to Cart