C12H316CEA36 Chemcia Scientific, LLC

1-(2-Amino-4-fluoro-phenyl)-ethanone

  • MDL number MFCD20621031
  • Cas Number : 159305-15-4
  • Molecular Weight : 153.15746

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2772

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2772
Availability details for
Quantity
  • Availability
  • Add to Cart