C12H316CEA48 Chemcia Scientific, LLC

3-Bromo-5-chloro-imidazo[1,2-a]pyridine

  • MDL number MFCD13193274
  • Cas Number : 1260656-94-7
  • Molecular Weight : 231.48033

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2789

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2789
Availability details for
Quantity
  • Availability
  • Add to Cart