C12H316CEA49 Chemcia Scientific, LLC

5-Chloro-3-iodo-imidazo[1,2-a]pyridine

  • MDL number MFCD27918591
  • Molecular Weight : 278.48073

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2790

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2790
Availability details for
Quantity
  • Availability
  • Add to Cart