C12H316CEA53 Chemcia Scientific, LLC

N-(5-Bromo-pyridin-2-yl)-4-methyl-benzenesulfonamide

  • MDL number MFCD00450557
  • Cas Number : 207801-52-3
  • Molecular Weight : 327.20167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2817

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2817
Availability details for
Quantity
  • Availability
  • Add to Cart