C12H316CEA6C Chemcia Scientific, LLC

(DL)-Isochroman-3-carboxylic acid

  • MDL number MFCD18416383
  • Cas Number : 1261578-13-5
  • Molecular Weight : 178.1894

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2799

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2799
Availability details for
Quantity
  • Availability
  • Add to Cart