C12H316CEA80 Chemcia Scientific, LLC

4-Chloro-2-fluoro-1-methoxy-benzene

  • MDL number MFCD00672964
  • Cas Number : 452-09-5
  • Molecular Weight : 160.57667

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Colorless Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3038

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3038
Availability details for
Quantity
  • Availability
  • Add to Cart