C12H316CEA85 Chemcia Scientific, LLC

2,7-Dibromo-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD22574163
  • Cas Number : 1380331-15-6
  • Molecular Weight : 276.91891

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3014

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3014
Availability details for
Quantity
  • Availability
  • Add to Cart