C12H316CEAA9 Chemcia Scientific, LLC

7-Chloro-imidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD09994935
  • Cas Number : 1020034-59-6
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloroimidazo[1,2-a]pyridine-2-carbaldehyde
Inchi Code 1S/C8H5ClN2O/c9-6-1-2-11-4-7(5-12)10-8(11)3-6/h1-5H
Inchi Key CIFSBBLEFJJPPY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3032

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3032
Availability details for
Quantity
  • Availability
  • Add to Cart