C12H316CEAE0 Chemcia Scientific, LLC

2-Chloromethyl-6-hydroxy-3H-quinazolin-4-one

  • MDL number MFCD22690476
  • Cas Number : 1258977-05-7
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2Z)-2-(chloromethylene)-6-hydroxy-2,3-dihydro-4(1H)-quinazolinone
Inchi Code 1S/C9H7ClN2O2/c10-4-8-11-7-2-1-5(13)3-6(7)9(14)12-8/h1-4,11,13H,(H,12,14)
Inchi Key XOCAPLOWSRISDB-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3150

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3150
Availability details for
Quantity
  • Availability
  • Add to Cart