C12H316CEAE2 Chemcia Scientific, LLC

2-Chloromethyl-quinazoline-4,6-diol

  • MDL number MFCD27918628
  • Cas Number : 1258977-05-7
  • Molecular Weight : 210.62134

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3152

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3152
Availability details for
Quantity
  • Availability
  • Add to Cart