C12H316CEAF8 Chemcia Scientific, LLC

2-Methoxy-4-methyl-5-nitro-benzoic acid

  • MDL number MFCD25968719
  • Cas Number : 90763-12-5
  • Molecular Weight : 211.17578

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3256

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3256
Availability details for
Quantity
  • Availability
  • Add to Cart