C12H316CEAFB Chemcia Scientific, LLC

1-Methoxymethoxy-3-nitro-benzene

  • MDL number MFCD27918631
  • Cas Number : 25458-48-4
  • Molecular Weight : 183.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(methoxymethoxy)-3-nitrobenzene
Inchi Code 1S/C8H9NO4/c1-12-6-13-8-4-2-3-7(5-8)9(10)11/h2-5H,6H2,1H3
Inchi Key GYQYQOZCWGUZBX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3267

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3267
Availability details for
Quantity
  • Availability
  • Add to Cart