C12H316CEAFE Chemcia Scientific, LLC

7-Chloro-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09991784
  • Cas Number : 1020038-42-9
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Inchi Code 1S/C8H5ClN2O2/c9-5-1-2-11-4-6(8(12)13)10-7(11)3-5/h1-4H,(H,12,13)
Inchi Key BEFUOITXMHRTRQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3275

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3275
Availability details for
Quantity
  • Availability
  • Add to Cart