C12H316CEB00 Chemcia Scientific, LLC

(4-Chloro-pyridin-2-yl)-dimethyl-amine

  • MDL number MFCD09031036
  • Cas Number : 735255-56-8
  • Molecular Weight : 156.61618

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3276

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3276
Availability details for
Quantity
  • Availability
  • Add to Cart