C12H316CEB01 Chemcia Scientific, LLC

(4-Chloro-pyridin-2-yl)-methyl-amine

  • MDL number MFCD08447077
  • Cas Number : 782439-26-3
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-N-methyl-2-pyridinamine
Inchi Code 1S/C6H7ClN2/c1-8-6-4-5(7)2-3-9-6/h2-4H,1H3,(H,8,9)
Inchi Key KCMQNXMUEQVQBM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3277

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3277
Availability details for
Quantity
  • Availability
  • Add to Cart