C12H316CEB05 Chemcia Scientific, LLC

1,4,6,7-Tetrahydro-pyrazolo[4,3-c]pyridine-3,5-dicarboxylic acid 5-tert-butyl ester

  • MDL number MFCD10697132
  • Cas Number : 518990-56-2
  • Molecular Weight : 267.28699

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3286

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3286
Availability details for
Quantity
  • Availability
  • Add to Cart