C12H316CEB0C Chemcia Scientific, LLC

5-Amino-2,4-dichloro-benzoic acid methyl ester

  • MDL number MFCD17276520
  • Cas Number : 84228-48-8
  • Molecular Weight : 220.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-amino-2,4-dichlorobenzoate
Inchi Code 1S/C8H7Cl2NO2/c1-13-8(12)4-2-7(11)6(10)3-5(4)9/h2-3H,11H2,1H3
Inchi Key WRCAZCJSGABHIT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3301

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3301
Availability details for
Quantity
  • Availability
  • Add to Cart