C12H316CEB19 Chemcia Scientific, LLC

4,5,6,7-Tetrahydro-2H-pyrazolo[4,3-c]pyridine-3-carboxylic acid ethyl ester hydrochloride

  • MDL number MFCD27997994
  • Cas Number : 1211512-51-4
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine-3-carboxylate hydrochloride
Inchi Code 1S/C9H13N3O2.ClH/c1-2-14-9(13)8-6-5-10-4-3-7(6)11-12-8;/h10H,2-5H2,1H3,(H,11,12);1H
Inchi Key MCDFHMGRGVFDOX-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HCl
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3278

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3278
Availability details for
Quantity
  • Availability
  • Add to Cart