C12H316CEB1A Chemcia Scientific, LLC

3-(4-Methoxy-benzyloxy)-phenylamine

  • MDL number MFCD09803253
  • Cas Number : 131770-85-9
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[(4-methoxybenzyl)oxy]aniline
Inchi Code 1S/C14H15NO2/c1-16-13-7-5-11(6-8-13)10-17-14-4-2-3-12(15)9-14/h2-9H,10,15H2,1H3
Inchi Key QEJXUWGQYJSOSA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Beige Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3302

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3302
Availability details for
Quantity
  • Availability
  • Add to Cart