C12H316CEB2D Chemcia Scientific, LLC

2,4,5,6-Tetrahydro-cyclopentapyrazole-3-carboxylic acid ethyl ester

  • MDL number MFCD13250144
  • Cas Number : 5932-31-0
  • Molecular Weight : 180.20819

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3375

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3375
Availability details for
Quantity
  • Availability
  • Add to Cart