C12H316CEB34 Chemcia Scientific, LLC

2-Chloro-6-methylamino-benzoic acid methyl ester

  • MDL number MFCD24387373
  • Cas Number : 1379595-97-7
  • Molecular Weight : 199.63855

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3384

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3384
Availability details for
Quantity
  • Availability
  • Add to Cart