C12H316CEB36 Chemcia Scientific, LLC

6-Chloro-2-ethyl-benzooxazole

  • MDL number MFCD21641331
  • Cas Number : 1027019-12-0
  • Molecular Weight : 181.62321

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Purple Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3386

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3386
Availability details for
Quantity
  • Availability
  • Add to Cart