C12H316CEB3A Chemcia Scientific, LLC

5-Chloro-2-dimethylamino-phenol

  • MDL number MFCD16998110
  • Cas Number : 57649-14-6
  • Molecular Weight : 171.628

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3391

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3391
Availability details for
Quantity
  • Availability
  • Add to Cart