C12H316CEB3D Chemcia Scientific, LLC

N,N-Bis-Boc-3-Nitro-pyridin-2-ylamine

  • MDL number MFCD28044131
  • Cas Number : 1044758-23-7
  • Molecular Weight : 339.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (tert-butoxycarbonyl)(3-nitropyridin-2-yl)carbamate
Inchi Code 1S/C15H21N3O6/c1-14(2,3)23-12(19)17(13(20)24-15(4,5)6)11-10(18(21)22)8-7-9-16-11/h7-9H,1-6H3
Inchi Key SDKVOBASBFCVML-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3402

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3402
Availability details for
Quantity
  • Availability
  • Add to Cart