C12H316CEB41 Chemcia Scientific, LLC

2-Methoxymethoxy-benzonitrile

  • MDL number MFCD18826226
  • Cas Number : 494209-23-3
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(methoxymethoxy)benzonitrile
Inchi Code 1S/C9H9NO2/c1-11-7-12-9-5-3-2-4-8(9)6-10/h2-5H,7H2,1H3
Inchi Key CAZBPZZSKGBVTE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3413

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3413
Availability details for
Quantity
  • Availability
  • Add to Cart